Learn More
Kartell™ Graduated Wide Mouth Square Bottles
Designed with a wide mouth for easy filling and emptying of liquid or powder. Kartell™ Polyethylene Graduated Wide Mouth Square Bottles provide a space saving design and are ideal for long term storage.
£133.00
Specifications
Material | Polyethylene |
---|---|
Certifications/Compliance | EC Regulation 1935/2004, EC Regulation 10/2011 |
Closure Included | Yes |
Autoclavable | Autoclavable |
Closure Material | Polypropylene |
Description
Designed with a wide mouth for easy filling and emptying of liquid or powder. Kartell™ Polyethylene Graduated Wide Mouth Square Bottles provide a space saving design and are ideal for long term storage.
- Approximate graduations allow reproducible filling without the use of cylinders/beaker
- Easy grip caps can be wired down for security
- Suitable for foodstuff in accordance with
- EC Regulation 1935/2004
- EC Regulation 10/2011
Spécification
Polyethylene | |
Yes | |
Polypropylene |
EC Regulation 1935/2004, EC Regulation 10/2011 | |
Autoclavable | |
Easy Grip |