Learn More
3M™ Graffiti Remover 1500
A safe and simple way to remove unsightly graffiti from plastic, painted, or varnished surfaces.
Brand:  3M™ 7000082043
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 17159340
  • £186.00 / 4 bottles
Estimated Shipment: 11-09-2024
to see stock.
Add to basket
Description
Description
  • Graffiti remover for plastic, painted or varnished surfaces
  • Effective against aerosol spray paint, permanent markers, lipstick, crayon, and emulsion paint
  • The solvent’s liquid form allows for quick penetration and rapid breakdown of graffiti
  • Does not contain NEP (N-Ethyl-pyrrolidone) or NMP (N-Methyl-2-pyrrolidone) used in other graffiti removers, which can pose a significant risk to health
  • Has minimal odor, enhancing user comfort and efficiency unlike traditional chemical removers which emit powerful odors and harmful vapors
  • Low environmental impact since the product does not contain aromatic, aliphatic, or chlorinated solvents
Specifications
Specifications
Graffiti Remover
Graffiti Removal
500 mL
Liquid
Plastic, Painted or Varnished Surfaces
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • 3M Graffiti Remover 1500
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-(2-Ethoxyethoxy)ethanol, benzyl alcohol, (2-Methoxymethylethoxy)propanol, Fatty acids, C16-18, 3-b