Learn More
3M™ Graffiti Remover 1500
A safe and simple way to remove unsightly graffiti from plastic, painted, or varnished surfaces.
Brand:  3M™ 7000082045
Additional Details : Weight : 20.00000kg
 View more versions of this product
Product Code. 17169340
  • £630.00 / 4 bottles
Estimated Shipment: 11-09-2024
to see stock.
Add to basket
Description
Description
  • Graffiti remover for plastic, painted or varnished surfaces
  • Effective against aerosol spray paint, permanent markers, lipstick, crayon, and emulsion paint
  • The solvent’s liquid form allows for quick penetration and rapid breakdown of graffiti
  • Does not contain NEP (N-Ethyl-pyrrolidone) or NMP (N-Methyl-2-pyrrolidone) used in other graffiti removers, which can pose a significant risk to health
  • Has minimal odor, enhancing user comfort and efficiency unlike traditional chemical removers which emit powerful odors and harmful vapors
  • Low environmental impact since the product does not contain aromatic, aliphatic, or chlorinated solvents
Specifications
Specifications
Graffiti Remover
Graffiti Removal
5 L
Liquid
Plastic, Painted or Varnished Surfaces