missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Gram Staining Kit
Use for convenient Gram staining of bacteria cells. Prolab Diagnostics™ Gram Staining Kit contains a collection of common stains.
£226.00 - £239.00
Specifications
Type | Gram Stain Kit |
---|---|
Certifications/Compliance | CE marked (98/78/EC) |
Format | Liquid |
For Use With (Application) | Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria |
Stain Type | Gram |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
12343378
|
Pro-Lab Diagnostics™
PL.8056/25 |
Crystal Violet 250mL, Grams Iodine 250mL, Grams Differentiator 250mL and Neutral Red 250mL |
£239.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12333378
|
Pro-Lab Diagnostics™
PL.8055/25 |
Crystal Violet 250mL, Grams Iodine 250mL, Grams Differentiator 250mL and Safranin 250mL |
£235.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12353378
|
Pro-Lab Diagnostics™
PL.8057/25 |
Crystal Violet 250mL, Grams Iodine 250mL, Grams Differentiator 250mL and Dilute Carbol Fuchsin 250mL |
£226.00
0.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions
- Solutions come in a selection of convenient and economical sizes.
- All stains are manufactured in accordance with all current regulatory requirements
- CE labelled
Specifications
Gram Stain Kit | |
Liquid | |
Gram |
CE marked (98/78/EC) | |
Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria | |
250 x 4 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Gram Staining Kit/Crystal Violet/Gram's Iodine/Gram's Differentiator/Neutral Red
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
- H371-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P273-Avoid release to the environment.
- P501-Dispose of contents/container to .
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P233-Keep container tightly closed.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : ethanol, C.I. basicviolet 3, methanol,ethanol, acetone, ethanol, methanol, ethanol, methanol, glyc
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pro-Lab Diagnostics™ Gram Staining Kit