missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Granulate Universal Coarse Grain

Granulate Universal Coarse Grain is ideal for setting oils, alkalis, lipids, fuels, lacquers etc.

Brand:  asecos™ 18468

 View more versions of this product

Product Code. 17896436

  • £158.00 / Pack of 3
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 1-3 mm grain size
  • 20 kg bag binds up to 16 L heating oil
  • Suitable for solid surfaces
  • Optimum slip resistance even when completely saturated
  • For indoors and outdoors
  • Available in one or three 20 kg bags.
Specifications

Specifications

Loose Particulate
Absorbent Granules
132.28 lb.
60 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.