missing translation for 'onlineSavingsMsg'
Learn More
Graphite flake, natural, -325 mesh, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11349278

Graphite flake, natural, -325 mesh, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11349278
250 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11349278

Brand: Thermo Scientific Alfa Aesar 043209.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Density ca. 2g/cm3
Mesh Size -325 Mesh
Assay Percent Range 99.8%, metal basis
Quantity 250 g
Physical Form Flakes
Chemical Name or Material Graphite, Natural

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM06L004 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number043209.A1,043209.18,043209.30
Lot NumberW13I019 Certificate TypeCertificate of Analysis Date18/07/2023 Catalog Number043209.A1,043209.18,043209.30
Lot NumberP03I046 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043209.A1,043224.22,043224.36,043209.18,043209.30
Lot NumberF01X009 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043209.A1,043224.22,043224.36,043209.18,043209.30
Lot NumberZ30B023 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043209.A1,043224.22,043224.36,043209.18,043209.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Graphite flake, natural, -325 mesh, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.