missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Graphite GC Liner O-Rings

Graphite liner O-rings for use with GC liners.

Brand:  Trajan™ 0726007

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15238863

  • £66.50 / Pack of 10
Estimated Shipment: 04-10-2024
to see stock.
Add to basket

Description

Description

  • Graphite Sealing Rings
  • Temperature stable up to 450°C
  • Supplied in packs of 10
Specifications

Specifications

Shimadzu 17 A (SPL-17 Injector)
450°C
10/Pk.
Graphite
Sealing Ring
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers