missing translation for 'onlineSavingsMsg'
Learn More
Graphite powder, synthetic, -20+100 mesh, 99% C, 0.2% max Ash, Thermo Scientific Chemicals Product Code.: 11305077

Graphite powder, synthetic, -20+100 mesh, 99% C, 0.2% max Ash, Thermo Scientific Chemicals

Product Code. 11305077
250 g, 250g
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11305077

Brand: Thermo Scientific Alfa Aesar 040769.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Graphite powder is basically used for casting, polishing purposes and molding. It is also used in pigments, anodes arc-lamp carbons, coating cathode ray tubes and moderator in nuclear piles. It is used in the preparation of single sheet graphene nanoflakes and graphite based magneto rheological elastomers. It is also employed as a free standing film like cantilever sensor in piezoelectric device and additionally used as a catalyst for wet peroxide oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Mesh Size −20+100 Mesh
Quantity 250 g
Solubility Information Insoluble in water.
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Graphite powder, Synthetic

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberT01K019 Certificate TypeCertificate of Analysis Date05/08/2024 Product Code040769.30,040769.A1
Lot NumberZ31H003 Certificate TypeCertificate of Analysis Date12/04/2022 Product Code040769.30,040769.A1
Lot NumberD30M08 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code040769.30,040769.A1
Lot NumberK29K12 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code040769.30,040769.A1
Lot NumberE27X031 Certificate TypeCertificate of Analysis Date29/11/2021 Product Code040769.30,040769.A1
compliance-icons
Product Identifier
  • Graphite powder, synthetic
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Graphite

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Graphite powder, synthetic, -20+100 mesh, 99% C, 0.2% max Ash, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.