missing translation for 'onlineSavingsMsg'
Learn More
Graphite, powder, Thermo Scientific Chemicals Product Code.: 10731335

Graphite, powder, Thermo Scientific Chemicals

Product Code. 10731335
100 g, Glass bottle
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10731335

Brand: Thermo Scientific Acros 385031000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 4575
Formula Weight 12.01
Physical Form Powder
Chemical Name or Material Graphite

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463201 Certificate TypeCertificate of Analysis Date27/05/2024 Catalog Number385030010,385031000
Lot NumberA0458159 Certificate TypeCertificate of Analysis Date23/11/2023 Catalog Number385030010,385031000
Lot NumberA0457587 Certificate TypeCertificate of Analysis Date08/11/2023 Catalog Number385030010,385031000
Lot NumberA0451757 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number385030010,385031000
Lot NumberA0451686 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number385030010,385031000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Graphite, powder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.