Learn More
Trajan Scientific and Medical™ Graphite/Vespel Ferrule
15% Graphite / 85% Vespel ferrules provide leak-free system connections, leading to improved chromatography for non-MS systems
Brand:  Trajan Scientific and Medical™ 072662
 View more versions of this product
Product Code. 15218893
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
15% Graphite / 85% Vespel ferrules provide leak-free system connections, leading to improved chromatography for non-MS systems
  • Composite of graphite and Vespel
  • Mechanically robust, hard material, long lifetime
  • Forms a strong grip with the capillary column
Specifications
Specifications
2-hole Ferrule
3.2 mm
Graphite, Vespel
0.4 mm
3.7 mm
1/16 in. Fittings
10 Pack