missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences Strep-Tactin™XT Superflow™ Column
Strep-Tactin™XT Superflow™ column is a gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
£76.20 - £372.24
Specifications
For Use With (Application) | Protein Purification |
---|---|
Type | Gravity Flow Column |
Product Code | Brand | Capacity (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Quantity | Price | Quantity & Availability | ||||
15543077
|
IBA Lifesciences
2-4011-005 |
5 x 0.2 mL | 5 Columns | ||||||
15553077
|
IBA Lifesciences
2-4012-001 |
1 x 1 mL | 1 Column | ||||||
15563077
|
IBA Lifesciences
2-4013-001 |
1 x 5 mL | 1 Column | ||||||
15573077
|
IBA Lifesciences
2-4014-001 |
1 x 10 mL | 1 Column | ||||||
Description
- Strep-Tactin™XT Superflow™ columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
- As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which leads to an outstanding purity of the target protein.
- In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™XT Superflow™ has a higher affinity for Strep-tag™ fusion proteins, which leads to an improved immobilization and purification.
- In addition, Strep-Tactin™XT Superflow™ allows the application of denaturing agents such as urea during the purification procedure.
- To reduce the waiting period during the sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
Protein Purification | |
Gravity Flow Column |
For Research Use Only.