missing translation for 'onlineSavingsMsg'
Learn More

Deutsch Neumann™ Grey Natural Rubber Stoppers

For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Grey Natural Rubber Stoppers are suitable for general use.

Brand:  Deutsch Neumann™ 1010108

 View more versions of this product

Product Code. 10405251

  • £0.25 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Acc. to DIN 12 871
  • Durometer 40 Shore A
  • Available in a wide range of sizes
Specifications

Specifications

20mm
Gray
12 mm
Tapered Stopper
1/Ea.
8 mm
Natural Rubber
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Deutsch Neumann™ Grey Natural Rubber Stoppers > Bottom Diameter: 8mm; Height: 20mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.