Learn More
Sartorius™ Biosart™ Monitor
Microbiological and analytical filtration units offering superior performance
Brand: Sartorius™ 16403-47-06----K
162.48 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
The system enables the user to select the optimal hardware and consumables for their needs in quality assurance. At the heart of the system is a stainless steel manifold designed to accommodate all types of filter holder and funnels. Please note: grey filters turn black when wet. Multi-branch system for routine colony or particle counting.
Specifications
Gray/White | |
48Pack | |
4 bags of 12 | |
Monitor |
Sterile | |
48Pack | |
48Pack |
Safety and Handling
- X48BIOSART 100 47MM NOIR 0,45µ QUA
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose
Your input is important to us. Please complete this form to provide feedback related to the content on this product.