Learn More
Macherey-Nagel™ Lysis Buffer VL, Macherey-Nagel™
A highly concentrated solution of chaotropic ions for quick, efficient lysis and creation of appropriate binding conditions during purification of nucleic acids using compatible kit (e.g. NucleoSnap™ DNA Plasma and NucleoSpin™ Virus).
Brand: Macherey-Nagel™ 740833.200
Specifications
50-01-1, 7732-18-5 | |
< 66% | |
Purification of DNA | |
4.5 to 5.2 | |
200 mL | |
1.18 g/cm3 |
0.5 | |
Colorless | |
Compatible kits (NucleoSnap DNA Plasma, NucleoSpin Virus) | |
Liquid | |
Odorless | |
Guanidine hydrochloride, Water |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Buffer VL
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash with water thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.