missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes

Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.

Brand:  Greiner Bio-One 450472

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 12671501

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.

 

  • Tubes feature closure that acts as a valve, allowing appropriate capillary or funnel to be inserted without removing the cap
  • Fit into a 13 × 75mm carrier tube to facilitate bar code labeling, centrifugation, and storage
  • Tubes with gel contain an inert barrier on the bottom that moves upward to form a stable barrier between the clot/cells and serum/plasma during centrifugation and transport
  • All tubes require MiniCollect funnel or capillary collection device (sold separately)

 

Specifications

Specifications

MiniCollect Capillary Blood Collection Tube
Valved Cap
Yes
Small volume blood collection
Serum Sep
0.8 mL
Gold
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers