missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes
Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
£49.33 - £535.00
Specifications
Product Type | MiniCollect Capillary Blood Collection Tube |
---|---|
Closure Included | Yes |
For Use With (Application) | Small volume blood collection |
Product Code | Brand | Additive | Closure Color | Capacity (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Additive | Closure Color | Capacity (Metric) | Price | Quantity & Availability | |||
13466778
|
Greiner Bio-One
450470 |
Serum | Red | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12671501
|
Greiner Bio-One
450472 |
Serum Sep | Gold | 0.8 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12500127
|
Greiner Bio-One
450474 |
K3 EDTA | Lavender | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Collect, transport and process capillary blood. Greiner Bio-One MiniCollect™ Capillary Blood Collection System Tubes are especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required.
- Tubes feature closure that acts as a valve, allowing appropriate capillary or funnel to be inserted without removing the cap
- Fit into a 13 × 75mm carrier tube to facilitate bar code labeling, centrifugation, and storage
- Tubes with gel contain an inert barrier on the bottom that moves upward to form a stable barrier between the clot/cells and serum/plasma during centrifugation and transport
- All tubes require MiniCollect funnel or capillary collection device (sold separately)
Specifications
MiniCollect Capillary Blood Collection Tube | |
Yes | |
Small volume blood collection |