missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK 269962, Tocris Bioscience™
Potent and selective ROCK inhibitor
£277.77 - £1050.23
Chemical Identifiers
CAS | 850664-21-0 |
---|---|
Molecular Formula | C29H30N8O5 |
Molecular Weight (g/mol) | 570.61 |
InChI Key | YOVNFNXUCOWYSG-UHFFFAOYSA-N |
Synonym | aminofurazanyl-azabenzimidazole 6n, n-3-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-1h-imidazo 4,5-c pyridin-6-yl oxy phenyl-4-2-morpholin-4-yl ethoxy benzamide, kinome_3843, d02zqs, n-3-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-1h-imidazo 4,5-c pyridin-6-yl oxy phenyl-4-2-4-morpholinyl ethoxy benzamide, n-3-2-4-amino-furazan-3-yl-1-ethyl-1h-imidazo 4,5-c pyridin-6-yl oxy phenyl-4-2-4-morpholinyl ethyl oxy benzamide, n-3-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethylimidazo 5,4-d pyridin-6-yl oxyphenyl-4-2-morpholin-4-ylethoxy benzamide |
PubChem CID | 16095342 |
IUPAC Name | N-[3-[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethylimidazo[4,5-c]pyridin-6-yl]oxyphenyl]-4-(2-morpholin-4-ylethoxy)benzamide |
SMILES | CCN1C2=CC(=NC=C2N=C1C3=NON=C3N)OC4=CC=CC(=C4)NC(=O)C5=CC=C(C=C5)OCCN6CCOCC6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18045732
|
Tocris Bioscience
4009/10 |
10 mg |
£277.77
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18076022
|
Tocris Bioscience
4009/50 |
50 mg |
£1050.23
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent Rho kinase (ROCK) inhibitor (IC50 values are 1.6 and 4 nM for recombinant human ROCK1 and ROCK2 respectively). Displays greater than 30-fold selectivity for ROCK against a panel of serine/threonine kinases. Induces vasorelaxation in preconstricted rat aorta (IC50 = 35 nM); lowers blood pressure in a rat model of hypertension.Chemical Identifiers
850664-21-0 | |
570.61 | |
aminofurazanyl-azabenzimidazole 6n, n-3-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-1h-imidazo 4,5-c pyridin-6-yl oxy phenyl-4-2-morpholin-4-yl ethoxy benzamide, kinome_3843, d02zqs, n-3-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-1h-imidazo 4,5-c pyridin-6-yl oxy phenyl-4-2-4-morpholinyl ethoxy benzamide, n-3-2-4-amino-furazan-3-yl-1-ethyl-1h-imidazo 4,5-c pyridin-6-yl oxy phenyl-4-2-4-morpholinyl ethyl oxy benzamide, n-3-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethylimidazo 5,4-d pyridin-6-yl oxyphenyl-4-2-morpholin-4-ylethoxy benzamide | |
N-[3-[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethylimidazo[4,5-c]pyridin-6-yl]oxyphenyl]-4-(2-morpholin-4-ylethoxy)benzamide |
C29H30N8O5 | |
YOVNFNXUCOWYSG-UHFFFAOYSA-N | |
16095342 | |
CCN1C2=CC(=NC=C2N=C1C3=NON=C3N)OC4=CC=CC(=C4)NC(=O)C5=CC=C(C=C5)OCCN6CCOCC6 |
Specifications
850664-21-0 | |
aminofurazanyl-azabenzimidazole 6n, n-3-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-1h-imidazo 4,5-c pyridin-6-yl oxy phenyl-4-2-morpholin-4-yl ethoxy benzamide, kinome_3843, d02zqs, n-3-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-1h-imidazo 4,5-c pyridin-6-yl oxy phenyl-4-2-4-morpholinyl ethoxy benzamide, n-3-2-4-amino-furazan-3-yl-1-ethyl-1h-imidazo 4,5-c pyridin-6-yl oxy phenyl-4-2-4-morpholinyl ethyl oxy benzamide, n-3-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethylimidazo 5,4-d pyridin-6-yl oxyphenyl-4-2-morpholin-4-ylethoxy benzamide | |
CCN1C2=CC(=NC=C2N=C1C3=NON=C3N)OC4=CC=CC(=C4)NC(=O)C5=CC=C(C=C5)OCCN6CCOCC6 | |
570.61 | |
570.6 | |
GSK 269962 |
C29H30N8O5 | |
YOVNFNXUCOWYSG-UHFFFAOYSA-N | |
N-[3-[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethylimidazo[4,5-c]pyridin-6-yl]oxyphenyl]-4-(2-morpholin-4-ylethoxy)benzamide | |
16095342 | |
>99% |
Safety and Handling
Recommended Storage : Store at +4°C