missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 319347A

Potent and selective IKKε inhibitor

Brand:  Tocris Bioscience™ 5672/10

Product Code. 18741348

  • £363.00 / 10mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GSK 319347A is a potent and selective IKKε inhibitor (pIC50 = 7.4). Exhibits >100-fold selectivity for IKKε over IKKα IKKβ, and a range of other kinases.
Specifications

Specifications

Inhibitor
862812-98-4
469.53g/mol
IkB Kinase Inhibitors
IkB Kinase
C22H19N3O5S2
Store at -20°C
10 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only