missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GST SpinTrap™
For rapid small-scale purification of GST-tagged proteins directly from clarified or unclarified cell lysates
Brand: Cytiva 28-9523-59
Code : GX
Additional Details : Weight : 1.00000kg
Description
- Preserves protein antigenicity and function
- Yields up to 500μg of protein per column
- Bed volume: 50μL
- Binding capacity/column: up to 500μg recombinant glutathione S-transferase (rGST)
- Ligand: glutathione
- Average particle size: 90μm
- Binding capacity/mL chromatography medium: >25mg horse liver GST/mL medium
- Matrix: 4% agarose
- pH working range: 4 to 13
Specifications
50 mL | |
GST SpinTrap columns are designed for rapid small-scale purification of GST-tagged proteins directly from clarified or unclarified cell lysates | |
4% Agarose | |
90 μm | |
For simple, small-scale purification of GST-tagged proteins and rapid expression screening by centrifugation | |
Glutathione |
Up to 500 μg recombinant glutathione S-transferase (rGST) | |
4 to 13 | |
4 to 30°C, PBS and 0,05% Kathon CG/ICP Biocide | |
50 x 50 μL | |
Spin Column | |
50 Columns |