missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GSTrap™ 4B Prepacked Columns: 7 x 25mm
Use for convenient, high capacity one-step purification of glutathione S-transferase (GST) tagged proteins. Cytiva Life Sciences™ GSTrap™ 4B Prepacked Columns: 7 x 25mm are prepacked with Glutathione Sepharose™ 4B
£277.00
Specifications
Bed Volume | 1 mL |
---|---|
Diameter (English) Inner | 0.27 in. |
Diameter (Metric) Inner | 7 mm |
Description | GSTrap 4B columns are prepacked Glutathione Sepharose 4B columns for convenient, high capacity one-step purification of glutathione S-transferase (GST) tagged proteins. |
Flow Rate | <4 mL/min. |
Description
- Prepacked with Glutathione Sepharose™ 4B media, featuring a glutathione ligand coupled to 4% agarose
- May be used with syringe, peristaltic pump or ¨A¨KTA design chromatography system
- Purifies GST-tagged proteins from bacterial lysates and other glutathione S-transferases or glutathione-dependent proteins
- Average particle size: 90μm
- 1 and 5mL columns
- Matrix: Highly cross-linked 4% agarose
- Binding capacity: >25mg horse liver GST/mL
- pH stability working range: 4 to 13
1mL columns
- Bed dimensions: 7 × 25mm
- Flow velocity: <4mL/min.
Specifications
1 mL | |
7 mm | |
<4 mL/min. | |
4 to 13 | |
4°C to 30°C, 20% Ethanol | |
GSTrap 4B columns are used for the purification of GST-tagged proteins from bacterial lysates and other glutathione S-tranferases or glutathione-dependent proteins. Mild elution conditions preserve protein antigenicity and function. | |
Glutathione |
0.27 in. | |
GSTrap 4B columns are prepacked Glutathione Sepharose 4B columns for convenient, high capacity one-step purification of glutathione S-transferase (GST) tagged proteins. | |
Polypropylene | |
4% Highly Cross-linked Agarose | |
5 bar (0.5 MPa, 70 psi) | |
Pre-packed Column |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Gstrap 4B
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol