missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GSTrap™ FF Columns: 7 x 25mm

Purifies GST-tagged proteins in one step

£144.00 - £295.00

Specifications

Bed Volume 1 mL
Diameter (English) Inner 0.27 in.
Diameter (Metric) Inner 7 mm
Description GSTrap FF columns are prepacked Glutathione Sepharose Fast Flow columns for fast, convenient, one-step purification of glutathione S-transferase (GST) tagged proteins.
Flow Rate <4 mL/min.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10646125
View Documents
Cytiva
17-5130-01
5 x 1 mL
£295.00
5mL
In Stock
to see stock.
Add to basket
 
10248704
View Documents
Cytiva
17-5130-02
2 x 1 mL
£144.00
Pack of 2
In Stock
to see stock.
Add to basket
 
Description

Description

  • Mild elution conditions preserve antigenicity and function of proteins extracted
  • Prepacked with Glutathione Sepharose™ media
  • Average particle size: 90μm
  • Bed dimensions: 7 × 25mm
  • Flow velocity: <4mL/min.
Media
  • Matrix: Highly cross-linked 4% agarose
  • Ligand: glutathione and 10-carbon linker arm
  • Binding capacity: >10mg/mL
  • pH stability working range: 3 to 12

Specifications

Specifications

1 mL
7 mm
<4 mL/min.
3 to 12
4°C to 30°C, 20% Ethanol
90 μm
Pre-packed Column
0.27 in.
GSTrap FF columns are prepacked Glutathione Sepharose Fast Flow columns for fast, convenient, one-step purification of glutathione S-transferase (GST) tagged proteins.
Polypropylene
4% Highly Cross-linked Agarose
5 bar (0.5 MPa, 70 psi)
For the purification of GST-tagged proteins from bacterial lysates and other glutathione S-tranferases or glutathione-dependent proteins and for preparation of cytoplasmic extract
Glutathione and 10 carbon linker arm
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • GSTrap 1 ml, 5 x 1 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.