missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, ≥99.5%, (Crystalline/Ultra Pure), MP Biomedicals

Forms clear, colorless solutions at 6.0 M concentration. O.D. 260 nm of a 6.0 M solution: <0.03

£49.10 - £2080.00

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11432002
View Documents
MP Biomedicals
0210569625
25 g
£49.10
25g
Please to purchase this item. Need a web account? Register with us today!
11442002
View Documents
MP Biomedicals
0210569680
100 g
£63.30
100g
Please to purchase this item. Need a web account? Register with us today!
11452002
View Documents
MP Biomedicals
0210569690
500 g
£206.00
500g
Please to purchase this item. Need a web account? Register with us today!
11462002
View Documents
MP Biomedicals
0210569691
1 kg
£396.00
1kg
Please to purchase this item. Need a web account? Register with us today!
11472002
View Documents
MP Biomedicals
0210569694
5 kg
£2080.00
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

50-01-1
95.53
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
CHEBI:32735
C(=N)(N)N.Cl
CH6ClN3
PJJJBBJSCAKJQF-UHFFFAOYSA-N
5742
guanidine;hydrochloride

Specifications

50-01-1
CH6ClN3
PJJJBBJSCAKJQF-UHFFFAOYSA-N
guanidine;hydrochloride
5742
95.5
Crystalline
≥99.5%
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
C(=N)(N)N.Cl
95.53
CHEBI:32735
Ultrapure
Guanidine Hydrochloride
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

Product Identifier
  • Guanidine Hydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Guanidine Hydrochloride

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.