missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride (Colorless-to-White Crystals), Fisher BioReagents™

For isolation of RNA from cell extracts

Brand:  Fisher Bioreagents BP178-500

120.03 GBP valid until 2024-12-31
Use promo code "23286" to get your promotional price.



Code : 32

Additional Details : CAS Number : 50-01-1 Weight : 0.55000kg


 View more versions of this product

Product Code. 10071503

  • £143.45 / 500g
Estimated Shipment: 27-09-2024
to see stock.

Description

Description

For isolation of RNA from cell extracts

Guanidine Hydrochloride is a strong protein-denaturing agent used in the isolation of RNA from cell extracts. RNase activity is inhibited in the presence of this reagent.
Specifications

Specifications

Guanidine Hydrochloride
≥99.5%
500 g
178°C
95.53
White
50-01-1
guanidine hydrochloride
15, 4597
CH6ClN3
Solid
≥99.5%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Guanidine hydrochloride, without anticaking agent

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents

Documents

Certificates
Special Offers

Special Offers