missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine Hydrochloride (Colorless-to-White Crystals), Fisher BioReagents™
For isolation of RNA from cell extracts
£143.45 - £195.00
Specifications
Chemical Name or Material | Guanidine Hydrochloride |
---|---|
CAS | 50-01-1 |
Assay Percent Range | ≥99.5% |
Synonym | guanidine hydrochloride |
Merck Index | 15, 4597 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10071503
|
Fisher Bioreagents
BP178-500 |
500 g |
£143.45
500g |
Estimated Shipment: 10-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10543325
|
Fisher Bioreagents
BP178-1 |
1 kg |
£195.00
1kg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
For isolation of RNA from cell extracts
Guanidine Hydrochloride is a strong protein-denaturing agent used in the isolation of RNA from cell extracts. RNase activity is inhibited in the presence of this reagent.Specifications
Guanidine Hydrochloride | |
≥99.5% | |
15, 4597 | |
CH6ClN3 | |
Solid | |
≥99.5% |
50-01-1 | |
guanidine hydrochloride | |
178°C | |
95.53 | |
White |