missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- High chemical resistance
- Good mechanical resistance and durability
- Antistatic raw material
- Flame retardant material
- Reusable, can be decontaminated

Specifications
Specifications
Color | Yellow, Gray |
Material | Neoprene |
Coating | Polyamide |
For Use With (Application) | Maintenance, industrial cleaning, handling, intervention on chemical accidents, decontamination, operations of alkylation |
Length (Metric) | 50 cm |
Style | Type PB(3) (partial body) sleeves - 50 cm |
Thickness | 360 gsm |
Certifications/Compliance | EC Category PPE: 3, EC Certificate: n° 00075/197/162/11/12/0856, EC Attestation: n° 00075/197/162/11/12/0856 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Honeywell™ Guardian Sleeves >
Spot an opportunity for improvement?Share a Content Correction