missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ Easy-Read™ Individually Calibrated Liquid-In-Glass Thermometers: Partial Immersion
GREENER_CHOICE

Individual Calibration Report as needed for many standard operating procedures

Brand:  H-B Instrument™ B60202-0000

 View more versions of this product

Product Code. 13049142

  • £287.00 / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Fill: Black spirit
  • Each thermometer is individually calibrated
  • Celsius-scale
  • Designed to be very easy to read
  • Replace mercury thermometers with the same accuracy without any hazards
  • Nontoxic, nonhazardous, biodegradable
  • NIST-Certified detailed, laminated report of calibration
  • Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read
  • Individually calibrated to NIST standards at specific temperature and supplied with NIST report of calibration stating corrections to one-tenth the smallest division
  • Contain an environmentally friendly non-mercury liquid that is non-toxic, non-hazardous, and fully biodegradable
  • In case of breakage the liquid can be cleaned up easily with soap and water

  • “Lead-free”, yellow back glass and black liquid provides easy readability and fewer reading errors
  • Contains non-toxic, biodegradable, Enviro-Safe™ liquid
  • EnviroKleen™ certified

Applications:

Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.

Certifications

Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST™ standards used for the calibration. Individually serialized and made in the USA.

Specifications

Specifications

0°, 37°, 56°C
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Partial Immersion
300 mm
Enviro-Safe™ Liquid
-10°C
±0.5°C
76 mm
Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration. Individually serialized and made in the USA
Precision
12 in.
Glass
70°C
-10°C to +70°C
0.5°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers