Learn More
Hafnium carbide, Gray powder, 99.5% (Metals basis excluding Zr), Thermo Scientific Chemicals
Description
It is used as an intermediate in chemical research, Pharmaceutical Intermediates,Organic Intermediates. Used in rocket nozzle, especially for re-entry space rocket nose cone.Due to its high hardness and melting point, HfC also widely use in hard alloy additives.Used in ceramic industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 3890°C |
Color | Gray |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.5% (Metals basis excluding Zr) |
Solubility Information | it is insoluble in water. |
Formula Weight | 190.54 |
Odor | Odorless |
Density | 12.2 g/cm3 |
Chemical Name or Material | Hafnium carbide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N14L048 | Certificate of Analysis | 17/02/2025 | 012567.03,012567.18,012567.09 |
R03K008 | Certificate of Analysis | 08/05/2024 | 012567.18,012567.09 |
R05J004 | Certificate of Analysis | 20/02/2024 | 012567.18,012567.09 |
P22I043 | Certificate of Analysis | 12/04/2022 | 012567.18,012567.09 |
T26D036 | Certificate of Analysis | 17/11/2021 | 012567.18,012567.09 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.