missing translation for 'onlineSavingsMsg'
Learn More
Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals Product Code.: 15495985

Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals

Product Code. 15495985
25 g
Click to view available options
Quantity:
2.5 g
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 15495985

Brand: Thermo Scientific Alfa Aesar 011834.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hafnium chloride tetrahydrofuran complex is an efficient catalyst for the Diels-Alder cycloadditions, Ziegler-Natta polymerization of alkenes. It is used as control rods in nuclear reactors and in manufacture of light bulb filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13499-05-3
Molecular Formula Cl4Hf
Molecular Weight (g/mol) 320.29
MDL Number MFCD00011034
InChI Key PDPJQWYGJJBYLF-UHFFFAOYSA-J
Synonym hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basisShow MoreShow Less
PubChem CID 37715
IUPAC Name tetrachlorohafnium
SMILES Cl[Hf](Cl)(Cl)Cl

Specifications

Melting Point 320° (sublimation)
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range >98% (Metals basis excluding Zr)
Solubility Information Soluble in water,methanol and acetone.
Formula Weight 320.3
Odor Irritating
Sensitivity Moisture sensitive
Chemical Name or Material Hafnium(IV) chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU15K031 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number011834.22,011834.88,011834.14
Lot NumberW14G063 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number011834.22,011834.88,011834.14
Lot NumberW08J011 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog Number011834.22,011834.88,011834.14
Lot NumberR24H064 Certificate TypeCertificate of Analysis Date07/07/2022 Catalog Number011834.22,011834.88,011834.14
Lot NumberS23G015 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011834.22,011834.88,011834.14
compliance-icons
Product Identifier
  • Hafnium(IV) chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Hafnium(IV) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.