Learn More
Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%, Thermo Scientific Chemicals
Description
Hafnium chloride tetrahydrofuran complex is an efficient catalyst for the Diels-Alder cycloadditions, Ziegler-Natta polymerization of alkenes. It is used as control rods in nuclear reactors and in manufacture of light bulb filaments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 320° (sublimation) |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | >98% (Metals basis excluding Zr) |
Solubility Information | Soluble in water,methanol and acetone. |
Formula Weight | 320.3 |
Odor | Irritating |
Sensitivity | Moisture sensitive |
Chemical Name or Material | Hafnium(IV) chloride |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U15K031 | Certificate of Analysis | 30/10/2024 | 011834.22,011834.88,011834.14 |
W14G063 | Certificate of Analysis | 15/10/2024 | 011834.22,011834.88,011834.14 |
W08J011 | Certificate of Analysis | 19/02/2024 | 011834.22,011834.88,011834.14 |
R24H064 | Certificate of Analysis | 07/07/2022 | 011834.22,011834.88,011834.14 |
S23G015 | Certificate of Analysis | 12/04/2022 | 011834.22,011834.88,011834.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.