Learn More
Hafnium(IV) oxide, Spectrographic Grade, 99.9% (metals basis excluding Zr), Zr typically <80ppm, Thermo Scientific Chemicals
Description
Hafnium(IV) oxide is used in optical coatings and in advanced metal-oxide-semiconductor devices. It is used as a gate insulator in field-effect transistors due to its high dielectric constant. It plays an important role as a possible candidate for resistive-switching memories, as a refractory material in the insulation of devices such as thermocouples and as a high-κ dielectric in DRAM capacitors. Also, used in the preparation of hafnium tetrachloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2774°C |
Physical Form | Powder |
Quantity | 10 g |
Merck Index | 14,4588 |
Solubility Information | Insoluble in water. |
Formula Weight | 210.49 |
Percent Purity | 99.9% |
Grade | Electrochemical |
Odor | Odorless |
Density | 9.68 g/mL |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S01B022 | Certificate of Analysis | 26/03/2025 | 011836.04,011836.09 |
L29K03 | Certificate of Analysis | 07/12/2021 | 011836.04,011836.09 |
L16M46 | Certificate of Analysis | 07/12/2021 | 011836.04,011836.09 |
S01B022 | Certificate of Analysis | 07/12/2021 | 011836.04,011836.09 |
Y25A035 | Certificate of Analysis | 24/11/2021 | 011836.04,011836.09 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.