missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Halt™ Phosphatase Inhibitor Single-Use Cocktail

Product Code. 10668304 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 mL
10 mL
24 x 100 μL
5 x 1 mL
Unit Size:
10mL
1mL
5mL
Each
This item is not returnable. View return policy

Product Code. 10668304

Brand: Thermo Scientific™ 78420

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Stabilize serine, threonine and tyrosine phosphorylation of proteins in cells and tissues with this cocktail of phosphatase inhibitors.

Preserve phosphorylation of proteins in cells and tissues with Thermo Scientific™ Halt Phosphatase Inhibitor Cocktail.

  • Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from serine/threonine phosphatases, as well as protein tyrosine phosphatases (PTPs) during cell lysis
  • The cocktail contains a mixture of four inhibitors of broad specificity including sodium fluoride, sodium orthovanadate, sodium pyrophosphate, and beta-glycerophosphate
  • Conveniently packaged in a 100X format, the cocktail is stable at 4°C and is suitable for use with any cell or tissue extract
  • The cocktail preserves protein phosphorylation in immunoprecipitation and kinase assays
  • Phosphorylation and dephosphorylation comprise a molecular on/off switch that regulates many key biological pathways within the cell including signal transduction, cell division, and apoptosis
  • To effectively study phosphorylation activation states, one must inactivate phosphatases to prevent their uncontrolled activity upon cell lysis

Additional Features

  • Devoid of protease inhibitors – To protect against both proteases and phosphatases, use Halt Protease and Phosphatase Inhibitor Cocktail (78420)
  • Dialyzable – Either dialyze or desalt sample to effectively remove inhibitors from cell extracts before performing procedures that are discovered to be incompatible with cocktail components
  • Supplied as a 100X concentrate – effective when used at a final concentration of 1X; however, samples that contain particularly high levels of phosphatase activity may require a final concentration of 2-3X
  • Compatibility – compatible with most detergent-based cell lysis reagents, including all Thermo Scientific Pierce Cell Lysis Solutions
  • All-in-one format – inhibits both serine-threonine phosphatases and protein tyrosine phosphatases
  • Compatible with popular protein assays – works with standard protein assays, including BCA and coomassie (Bradford) assays
  • Convenient storage – refrigerator storage (2 to 8°C) saves valuable space in the freezer and allows for immediate use
TRUSTED_SUSTAINABILITY

Specifications

Product Type Phosphatase Inhibitor Cocktail
Content And Storage Upon receipt store product at 4°C. Do not freeze.
Form Liquid
Product Line Halt
Reagent Type Phosphatase Inhibitor
Inhibitors Phosphatase Inhibitor
Quantity 1 mL
compliance-icons
Product Identifier
  • Halt Phosphatase Inhibitor Cocktail
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : sodium fluoride, trisodium tetraoxovanadate

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.