missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Halt™ Phosphatase Inhibitor Single-Use Cocktail

Product Code. 11824101 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 mL
10 mL
24 x 100 μL
5 x 1 mL
Unit Size:
10mL
1mL
5mL
Each
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11824101

Brand: Thermo Scientific™ 78427

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy
Regler: Denna produkt innehåller ett ämne som är upptaget i bilaga XVII till REACH-förordningen (EG) nr 1907/2006. Alla kunder måste fylla i en deklaration före leverans.

Stabilize serine, threonine and tyrosine phosphorylation of proteins in cells and tissues with this cocktail of phosphatase inhibitors.

Preserve phosphorylation of proteins in cells and tissues with Thermo Scientific™ Halt Phosphatase Inhibitor Cocktail.

  • Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from serine/threonine phosphatases, as well as protein tyrosine phosphatases (PTPs) during cell lysis
  • The cocktail contains a mixture of four inhibitors of broad specificity including sodium fluoride, sodium orthovanadate, sodium pyrophosphate, and beta-glycerophosphate
  • Conveniently packaged in a 100X format, the cocktail is stable at 4°C and is suitable for use with any cell or tissue extract
  • The cocktail preserves protein phosphorylation in immunoprecipitation and kinase assays
  • Phosphorylation and dephosphorylation comprise a molecular on/off switch that regulates many key biological pathways within the cell including signal transduction, cell division, and apoptosis
  • To effectively study phosphorylation activation states, one must inactivate phosphatases to prevent their uncontrolled activity upon cell lysis

Additional Features

  • Devoid of protease inhibitors – To protect against both proteases and phosphatases, use Halt Protease and Phosphatase Inhibitor Cocktail (78420)
  • Dialyzable – Either dialyze or desalt sample to effectively remove inhibitors from cell extracts before performing procedures that are discovered to be incompatible with cocktail components
  • Supplied as a 100X concentrate – effective when used at a final concentration of 1X; however, samples that contain particularly high levels of phosphatase activity may require a final concentration of 2-3X
  • Compatibility – compatible with most detergent-based cell lysis reagents, including all Thermo Scientific Pierce Cell Lysis Solutions
  • All-in-one format – inhibits both serine-threonine phosphatases and protein tyrosine phosphatases
  • Compatible with popular protein assays – works with standard protein assays, including BCA and coomassie (Bradford) assays
  • Convenient storage – refrigerator storage (2 to 8°C) saves valuable space in the freezer and allows for immediate use
TRUSTED_SUSTAINABILITY

Specifikationer

Product Type Phosphatase Inhibitor Cocktail
Content And Storage Upon receipt store product at 4°C. Do not freeze.
Form Liquid
Product Line Halt
Reagent Type Phosphatase Inhibitor
Inhibitors Phosphatase Inhibitor
Quantity 10 mL
compliance-icons
Produktbeteckning
  • Halt Phosphatase Inhibitor Cocktail
Signalord
  • Warning
Riskkategorier
  • Acute toxicity Category 4
Faroangivelser
  • H302-Harmful if swallowed.
Skyddsangivelse
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Kompletterande information
  • MIXTURE LIST-Contains : sodium fluoride, trisodium tetraoxovanadate

For Research Use Only. Not for use in diagnostic procedures.

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.