Learn More
Thermo Scientific™ Halt™ Protease and Phosphatase Inhibitor Cocktail (100X)
Shop All Thermo Scientific ProductsDescription
The all-in-one Thermo Scientific™ Halt Protease and Phosphatase Inhibitor Cocktail provides the convenience of a single solution with full sample protection.
Additional Information and Online Ordering
Highlights:
Complete protection all-in-one cocktail contains both protease and phosphatase inhibitors
Multiple package sizes choose from four convenient packages sizes to match nearly any scale
Convenient the 100X liquid formulation is much easier and more effective to use than individual inhibitors and other formats
Easy storage aqueous-based format allows for convenient cold-room storage without freezing, by contrast with DMSO-based cocktails
Compatible can be used in mass spectrometry (MS) because it does not contain AEBSF, which can cause peaks to shift
Validated fully compatible with Thermo Scientific Pierce™ Cell Lysis Buffers
Includes:
Formulations with and without supplementary vial of EDTA for metalloprotease inhibition, both of which are offered in four package sizes.
Specifications
Specifications
| Product Type | Protease and Phosphatase Inhibitor Cocktail |
| Content And Storage | Upon receipt store at 4°C |
| Form | Liquid |
| Product Line | Halt |
| Buffer | Lysis Buffer |
| Reagent Type | Protease Inhibitor, Phosphatase Inhibitor |
| Inhibitors | Protease Inhibitor, Phosphatase Inhibitor |
| Quantity | 1 mL |
Safety and Handling
- Halt Protease and Phosphatase Inhibitor Cocktail (100X)
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P270-Do not eat, drink or smoke when using this product.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : sodium fluoride, trisodium tetraoxovanadate
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.