missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Halt™ Protease and Phosphatase Inhibitor Cocktail (100X)

Product Code. p-7184459 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 mL
10 mL
24 x 100 μL
5 x 1 mL
Unit Size:
10mL
1mL
2400µL
Pack of 5
This item is not returnable. View return policy

Product Code. 10025743

Brand: Thermo Scientific™ 78442

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prevent protein degradation and stabilize phosphorylation with this all-in-one cocktail of protease and phosphatase inhibitors.

The all-in-one Thermo Scientific™ Halt Protease and Phosphatase Inhibitor Cocktail provides the convenience of a single solution with full sample protection.

Additional Information and Online Ordering

This broad-spectrum Halt Protease and Phosphatase Inhibitor Cocktail is specifically optimized to protect proteins from degradation during extraction and purification. The cocktail contains inhibitors against the major classes of proteases and phosphatases, targeting aminopeptidases, cysteine and serine proteases and serine/threonine and protein tyrosine phosphatases. The 100X cocktails, available in regular and EDTA-free formulations and several package formats, are refrigerator-stable liquids that are very easy to use for nearly any sample or experimental scale.

Highlights:

  • Complete protection all-in-one cocktail contains both protease and phosphatase inhibitors

  • Multiple package sizes choose from four convenient packages sizes to match nearly any scale

  • Convenient the 100X liquid formulation is much easier and more effective to use than individual inhibitors and other formats

  • Easy storage aqueous-based format allows for convenient cold-room storage without freezing, by contrast with DMSO-based cocktails

  • Compatible can be used in mass spectrometry (MS) because it does not contain AEBSF, which can cause peaks to shift

  • Validated fully compatible with Thermo Scientific Pierce™ Cell Lysis Buffers

Includes:

Formulations with and without supplementary vial of EDTA for metalloprotease inhibition, both of which are offered in four package sizes.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Protease and Phosphatase Inhibitor Cocktail
Content And Storage Upon receipt store at 4°C
Form Liquid
Product Line Halt
Buffer Lysis Buffer
Reagent Type Protease Inhibitor, Phosphatase Inhibitor
Inhibitors Protease Inhibitor, Phosphatase Inhibitor
Quantity 24 x 100 μL
compliance-icons
Product Identifier
  • HAlt protease
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : trisodium orthovanadate, sodium fluoride

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.