Learn More
Thermo Scientific™ Halt™ Protease and Phosphatase Inhibitor Cocktail, EDTA-free (100X)
Prevent protein degradation and stabilize phosphorylation with this all-in-one cocktail of protease and phosphatase inhibitors.
Brand: Thermo Scientific™ 78445
431.68 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
The all-in-one Thermo Scientific™ Halt Protease and Phosphatase Inhibitor Cocktail provides the convenience of a single solution with full sample protection.
Additional Information and Online Ordering
This broad-spectrum Halt Protease and Phosphatase Inhibitor Cocktail is specifically optimized to protect proteins from degradation during extraction and purification. The cocktail contains inhibitors against the major classes of proteases and phosphatases, targeting aminopeptidases, cysteine and serine proteases and serine/threonine and protein tyrosine phosphatases. The 100X cocktails, available in regular and EDTA-free formulations and several package formats, are refrigerator-stable liquids that are very easy to use for nearly any sample or experimental scale.
Highlights:
- Complete protection – all-in-one cocktail contains both protease and phosphatase inhibitors
- Multiple package sizes – choose from four convenient packages sizes to match nearly any scale
- Convenient – the 100X liquid formulation is much easier and more effective to use than individual inhibitors and other formats
- Easy storage – aqueous-based format allows for convenient cold-room storage without freezing, by contrast with DMSO-based cocktails
- Compatible – can be used in mass spectrometry (MS) because it does not contain AEBSF, which can cause peaks to shift
- Validated – fully compatible with Thermo Scientific Pierce™ Cell Lysis Buffers
Includes:
Formulations with and without supplementary vial of EDTA for metalloprotease inhibition, both of which are offered in four package sizes.
Storage Requirements:
Upon receipt store at 4°C. Do not freeze product.
Specifications
Halt Protease and Phosphatase Inhibitor Cocktail, EDTA-Free | |
Upon receipt store at 4°C. Product shipped with an ice pack. Do not freeze product. | |
Halt™ | |
Protease Inhibitor, Phosphatase Inhibitor | |
5 x 1 mL |
Protease and Phosphatase Inhibitor Cocktail | |
Liquid | |
Lysis Buffer | |
Protease Inhibitor, Phosphatase Inhibitor |
Safety and Handling
- Halt Protease and Phosphatase Inhibitor Cocktail, EDTA-free (100X)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium fluoride, trisodium tetraoxovanadate
For Research Use Only. Not for use in diagnostic procedures.