missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Ham's F-10 Nutrient Mix
Ham's F-10 Nutrient Mix
£6.78 - £165.00
Specifications
Content And Storage | 2 to 8°C |
---|---|
Cell Line | CHO |
Concentration | 1 X |
Form | Liquid |
Product Type | Ham's F-10 Nutrient Mix |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11554436
|
Gibco™
31550015 |
100 mL |
£6.78
100mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11564436
|
Gibco™
31550023 |
500 mL |
£18.77
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11584436
|
Gibco™
31550049 |
20 x 100 mL |
£100.00
2000mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11574436
|
Gibco™
31550031 |
10 x 500 mL |
£165.00
Pack of 10 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells. F-10 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including COS-7, primary rat astrocytes, and rat prostate epithelial cells.Compared to other basal media, F-10 contains a wider variety of components, including zinc, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-10 has led to a variety of improved formulations, such as Ham's F-12 Nutrient Mixture (F-12).
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ F-10 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ F-10 product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Gibco™ F-10 contains no proteins or growth factors. Therefore, F-10 often requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-10 uses a sodium bicarbonate buffer system (1.2 g/L), and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
2 to 8°C | |
1 X | |
Ham's F-10 Nutrient Mix | |
No HEPES | |
7.0 to 7.4 |
CHO | |
Liquid | |
Glutamine, Phenol Red, Sodium Pyruvate | |
Mammalian cell culture |