missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Ham's F-10 Nutrient Mix, GlutaMAX™ Supplement
Ham's F-10 Nutrient Mix, GlutaMAX™ Supplement
Brand: Gibco™ 41550021
Code : 50
Additional Details : Weight : 0.52500kg
Description
Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells. F-10 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including COS-7, primary rat astrocytes, and rat prostate epithelial cells.Compared to other basal media, F-10 contains a wider variety of components, including zinc, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-10 has led to a variety of improved formulations, such as Ham's F-12 Nutrient Mixture (F-12).
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ F-10 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
Gibco™ F-10 contains no proteins or growth factors. Therefore, F-10 often requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-10 uses a sodium bicarbonate buffer system (1.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
Ham's F-10 Nutrient Mix, 500mL, 1 X Concentration, Liquid, 7.2 to 7.4 pH, 12 Months Shelf Life, 275 to 315 mOsm/kg Osmolality, With Sodium Pyruvate, GlutaMAX™, Phenol Red, 2 to 8°C Storage, For Mammalian Cell Culture | |
1 X | |
Ham's F-10 Nutrient Mix | |
No HEPES | |
7.2 to 7.4 |
2 to 8°C | |
CHO | |
Liquid | |
GlutaMAX, Phenol Red, Sodium Pyruvate | |
Mammalian cell culture | |
500 mL |