missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Ham's F-10 Nutrient Mix, GlutaMAX™ Supplement
Ham's F-10 Nutrient Mix, GlutaMAX™ Supplement
£26.35 - £226.00
Specifications
Certifications/Compliance | ISO 13485 standards, FDA |
---|---|
Content And Storage | 2 to 8°C |
Cell Line | CHO |
Concentration | 1 X |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11554486
|
Gibco™
41550021 |
500 mL |
£26.35
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11564486
|
Gibco™
41550088 |
10 x 500 mL |
£226.00
Pack of 10 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells. F-10 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including COS-7, primary rat astrocytes, and rat prostate epithelial cells.Compared to other basal media, F-10 contains a wider variety of components, including zinc, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-10 has led to a variety of improved formulations, such as Ham's F-12 Nutrient Mixture (F-12).
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ F-10 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
Gibco™ F-10 contains no proteins or growth factors. Therefore, F-10 often requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-10 uses a sodium bicarbonate buffer system (1.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
CHO | |
Liquid | |
GlutaMAX, Phenol Red, Sodium Pyruvate | |
Mammalian cell culture |
2 to 8°C | |
1 X | |
Ham's F-10 Nutrient Mix | |
No HEPES | |
7.2 to 7.4 |