Learn More
Hamilton™ Syringe Guide for Microliter™ and Gaslight™ Syringes
Designed to work with 700/7000 series MICROLITER syringes and 1700 series GASTIGHT syringes
Brand:  Hamilton™ 14906
Code : 13
Additional Details : Weight : 0.01700kg
 View more versions of this product
Product Code. 10606473
  • £75.35 / Each
Estimated Shipment: 07-08-2024
to see stock.
Description
Description
Designed to work with 700/7000 series MICROLITER syringes and 1700 series GASTIGHT syringes
Syringe guides are designed to prevent plunger bending and the adjustable stop prevents plunger damage.
Specifications
Specifications
Syringe Guide
Microliter and Gastight Syringes, 7000 Series and 25 to 500μL Syringes