Learn More
Hamilton™ Syringe Guide for Microliter™ and Gaslight™ Syringes
Designed to work with 700/7000 series MICROLITER syringes and 1700 series GASTIGHT syringes
£68.06 - £75.35
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10130221
|
Hamilton™
14806 |
Microliter and Gastight Syringes and 700 and 1700 Syringe Series |
£68.06
Each |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10606473
|
Hamilton™
14906 |
Microliter and Gastight Syringes, 7000 Series and 25 to 500μL Syringes |
£75.35
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Designed to work with 700/7000 series MICROLITER syringes and 1700 series GASTIGHT syringes
Syringe guides are designed to prevent plunger bending and the adjustable stop prevents plunger damage.
Specifications
Syringe Guide |