missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kleenex™ Fragrance Free Frequent Use Hand Cleanser

Ideal for dispensers in busy washrooms. Kleenex™ Fragrance Free Frequent Use Hand Cleanser is a colorless liquid to help maintain hygiene and cleanliness.

Brand:  Kimberly-Clark™ 6333

62.49 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 12422320

  • £147.00 / Pack of 6
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Supplied in packs of 6 x 1 L refill cassettes
  • Clear and fragrance-free to suit all skin types
  • Consistent shot size
  • Contains up to 1,000 shots per cassette
  • Quick and easy to install
  • Recyclable when empty
  • Compatible with Aquarius™ and Kimberly-Clark Professional™ Hand Cleanser Dispensers
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hand Cleanser
Liquid
6
33.8 oz.
13.6 lb.
Clear
93 x 93 x 180 mm
6 x 1 L
1000 mL
1.033 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark™ Kleenex™ Fragrance Free Frequent Use Hand Cleanser > Volume: 1000 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.