missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Hand Operated Solvent Pump for Tin-foil Canisters

Buerkle™ Hand Operated Solvent Pump for Tin-foil Canisters remove contamination of organic solvents and flavouring agents.

Brand:  Buerkle™ 5603-2000

Product Code. 10482594

  • £566.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Medium-contacting parts made of stainless steel AISI 304 (1.4301) and PTFE
  • Rigid discharge tube with stopcock made of stainless steel/PTFE
  • Suitable for containers with R 2 in. thread
  • Thread adapter for commercially available containers available as accessory
Specifications

Specifications

Solvents
60 cm
Stainless Steel, PTFE
For pull-out spout with 35 mm diameter
Thread Adapter
Explosion Categories IIA, IIB and IIC, for use in Ex protection zone 0 and 1
Tin-Foil Canister
Hand Operated Solvent Pump
10 L/min.
2.052 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ Hand Operated Solvent Pump for Tin-foil Canisters > Flow Rate: 10LPM

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.