Learn More
Buerkle™ OTAL™ Hand Pump
Ensure removal of residual liquid with these versatile hand pumps. Buerkle™ OTAL™ Hand Pumps feature a telescopic immersion tube that adapts itself to the container height on insertion, allowing the compact, space-saving hand pump to be used with different containers.
Brand: Buerkle™ 5005-6000
79.26 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : A0
Additional Details : Weight : 0.67500kg
Alert:
Attention! Make sure to pay attention to the “List of Chemical Resistance“!
Description
OTAL hand pumps fit most commercial storage drums. The telescopic immersion tube adapts itself to the container height on insertion. This ensures good removal of residual liquid and means that the hand pump can be used with different containers.- Particularly compact, space-saving hand pump
- Good pump capacity
- With universal bungs made of soft PVC, for container openings of 30–70 mm (1.18-2.76 in.) Ø
- resistant to weak acids, alkalis and detergents.
- particularly suitable for acids and alkalis, not for heavily oxidizing acids and alkalis.
- With its special material properties, PVDF has an even better chemical resistance than PP. Resistant to chlorine bleach.
- for liquids that are not easily combustible in Hazard Category AIII (VbF).
- The OTAL PP or PVDF hand pump should be used for caustic media.
- discharge hose with a length of 1.5 m made of PVC and PP/PE stopcock, delivery tube made of PP.
- The discharge hose is particularly suited to fill distant containers.
Specifications
70 cm | |
Hand Pump | |
Alkalis, Detergent, Acid | |
Storage Drum | |
12 L/min. | |
0.58 kg |
Co mmercial Storage | |
70 cm | |
Polypropylene, PVC | |
30 to 70 mm | |
12 mm Dia. Polypropylene/PVC Delivery Tube, Tap with Discharge Hose |