Learn More
Brady™ HandiMark™ Vinyl Repositionable Film
Brand:  Brady™ 042103
 View more versions of this product
Product Code. 12810600
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
HandiMark Repositionable Vinyl Film
Yellow
Vinyl
For temporary, removable and repositionable labels, like 5S and lean inventory labels
1 Roll
0.031 mm
13 mm
Acrylic, Repositionable
Gloss
Repositionable Film
HandiMark
-40°C to +82°C
15 m
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ HandiMark™ Vinyl Repositionable Film > Color: White; Dimensions (LxW): 15m x 13mm