Learn More
Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Nonsterile
GREENER_CHOICE
Black tips for accurate sample delivery
Brand:  Axygen™ TT-20-CBK-HTR
 View more versions of this product
Product Code. 11314295
  • £300.77 / Pack of 2304
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polypropylene
  • Volume ranging from 20 to 1000μL
  • Available with or without aerosol-resistant filters
  • Compatible with Axygen™ Tecan™/Perkin Elmer/Auroroa™ Biomed/Corbett Cas-1200 systems
Certifications
Certified DNAse-, RNAse- and Pyrogen-safe
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 μL
Non-sterile
Autoclavable
For accurate sample delivery
Automation, Round
Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head
Non-filtered
Black
Non-ANSI
Rack
Hanging Tip Racked
Polypropylene