Learn More
Faster™ Hard Exhaust Duct Connection for SafeFAST and CytoFAST Cabinets
Hard duct connection on the top of the cabinet to connect to ductwork.
Brand:  Faster™ FX0002094040
Code : NEW
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15513366
  • £356.00 / Each
Estimated Shipment: 25-11-2024
to see stock.
Description
Description
  • 200 mm diameter
  • Specifically for SafeFAST or CytoFAST cabinets
Specifications
Specifications
Ducting Safety Cabinets
SafeFAST Classic, Elite, Premium, CytoFAST Elite 209