Learn More
Faster™ Hard Exhaust Duct Connection for SafeFAST and CytoFAST Cabinets
Hard duct connection on the top of the cabinet to connect to ductwork.
£360.00 - £443.00
Specifications
For Use With (Application) Ducting Safety Cabinets
View More Specs
Products 4
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
16268590
View Documents
Faster™
FX0002154040
SafeFAST Classic, Elite, Premium, 109, 215, 315
£427.00
Each
15513366
View Documents
Faster™
FX0002094040
SafeFAST Classic, Elite, Premium, CytoFAST Elite 209
£360.00
Each
15553366
View Documents
Faster™
FX0002124040
SafeFAST Classic, Elite, Premium, CytoFAST Elite 212/312
£392.00
Each
15573376
View Documents
Faster™
FX0002184040
SafeFAST Classic, Elite, Premium, CytoFAST Elite 112, 115, 218, 318
£443.00
Each
Description
Description
  • 200 mm diameter
  • Specifically for SafeFAST or CytoFAST cabinets
Specifications
Specifications
Ducting Safety Cabinets