Learn More
Faster™ Hard Exhaust Duct Connection for SafeFAST and CytoFAST Cabinets
Hard duct connection on the top of the cabinet to connect to ductwork.
Brand: Faster™ FX0002124040
This item is not returnable.
View return policy
Description
- 200 mm diameter
- Specifically for SafeFAST or CytoFAST cabinets
Specifications
Ducting Safety Cabinets | |
SafeFAST Classic, Elite, Premium, CytoFAST Elite 212/312 |