Learn More
Faster™ Hard Exhaust Duct Connection for SafeFAST and CytoFAST Cabinets
Hard duct connection on the top of the cabinet to connect to ductwork.
Brand:  Faster™ FX0002184040
Code : NEW
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15573376
  • £438.00 / Each
Estimated Shipment: 25-11-2024
to see stock.
Description
Description
  • 200 mm diameter
  • Specifically for SafeFAST or CytoFAST cabinets
Specifications
Specifications
Ducting Safety Cabinets
SafeFAST Classic, Elite, Premium, CytoFAST Elite 112, 115, 218, 318