missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Ultimate Security Padlock

The 7 pin disk cylinder gives ultimate protection against manipulation and key interchanges.

Brand:  Brady™ 805838

Code : 50

Additional Details : Weight : 0.40000kg

 View more versions of this product

Product Code. 11701296

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Shackle and body are made of highest standard hardened alloy steel
Specifications

Specifications

Ultimate Security Padlock
Hardened Alloy Steel
11 mm
109 mm
Hardened Alloy Steel
1 Ea.
Black
Ultimate Security Padlock
63.00 x 109.00 mm
50.00 mm
7
63 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers